accessibility menu, dialog, popup

UserName

CAS RN 35320-23-1

IUPAC Name: 2-aminopropan-1-ol
Synonyms: 2-aminopropanol
Molecular Weight (g/mol): 75.11
Molecular Formula: C3H9NO
InChi Key: BKMMTJMQCTUHRP-UHFFFAOYNA-N
SMILES: CC(N)CO
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)