accessibility menu, dialog, popup

UserName

FFFI

CAS RN 353-83-3

IUPAC Name: 1,1,1-trifluoro-2-iodoethane
Synonyms: 1,1,1-trifluoro-2-iodoethane
Molecular Weight (g/mol): 209.94
Molecular Formula: C2H2F3I
InChi Key: RKOUFQLNMRAACI-UHFFFAOYSA-N
SMILES: FC(F)(F)CI
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)