accessibility menu, dialog, popup

UserName

CAS RN 353-42-4

BFFFOH3CH3C

CAS RN 353-42-4

IUPAC Name: (dimethyloxo)trifluoro-λ⁵-borane
Synonyms: (dimethyloxo)trifluoro-λ⁵-borane
Molecular Weight (g/mol): 113.87
Molecular Formula: C2H6BF3O
InChi Key: FOWZWMGLMYVYDH-UHFFFAOYSA-N
SMILES: C[O+](C)[B-](F)(F)F
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)