accessibility menu, dialog, popup

UserName

CAS RN 3528-17-4

OS

CAS RN 3528-17-4

IUPAC Name: 3,4-dihydro-2H-1-benzothiopyran-4-one
Synonyms: 2,3-dihydro-1-benzothiopyran-4-one
Molecular Weight (g/mol): 164.22
Molecular Formula: C9H8OS
InChi Key: CVQSWZMJOGOPAV-UHFFFAOYSA-N
SMILES: O=C1CCSC2=CC=CC=C12
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results