accessibility menu, dialog, popup

UserName

CAS RN 352195-40-5

N(R)N(R)ONP(R)(S)HNOOOOOOHHONaNa

CAS RN 352195-40-5

IUPAC Name: disodium hydrate [3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methyl phosphate
Synonyms: disodium hydrate [3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl phosphate
Molecular Weight (g/mol): 410.19
Molecular Formula: C10H13N4Na2O9P
InChi Key: KQCIGEIXDXQSGM-UHFFFAOYNA-L
SMILES: O.[Na+].[Na+].OC1C(COP([O-])([O-])=O)OC(C1O)N1C=NC2=C1NC=NC2=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (6)