accessibility menu, dialog, popup

UserName

CAS RN 352018-92-9

ONI

CAS RN 352018-92-9

IUPAC Name: 5-iodo-2-phenoxypyridine
Synonyms: 5-iodo-2-phenoxypyridine
Molecular Weight (g/mol): 297.1
Molecular Formula: C11H8INO
InChi Key: JCEXQQNAXWYXRX-UHFFFAOYSA-N
SMILES: IC1=CN=C(OC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results