accessibility menu, dialog, popup

UserName

CH3SN

CAS RN 35120-10-6

IUPAC Name: 2-(methylsulfanyl)acetonitrile
Synonyms: 2-(methylsulfanyl)acetonitrile
Molecular Weight (g/mol): 87.14
Molecular Formula: C3H5NS
InChi Key: ZRIGDBVSVFSVLL-UHFFFAOYSA-N
SMILES: CSCC#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)