CAS RN 35090-64-3
IUPAC Name:
4-bromo-2-(hydroxymethyl)-6-methoxyphenol
Synonyms:
4-bromo-2-(hydroxymethyl)-6-methoxyphenol
Molecular Weight (g/mol):
233.06
Molecular Formula:
C8H9BrO3
InChi Key:
UBLQIISPEAEAIX-UHFFFAOYSA-N
SMILES:
COC1=C(O)C(CO)=CC(Br)=C1
1
–
1
of
1
results
1
Alfa Aesar™ 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol
CAS: 35090-64-3 Molecular Formula: C8H9BrO3 Molecular Weight (g/mol): 233.06 MDL Number: MFCD02081986 InChI Key: UBLQIISPEAEAIX-UHFFFAOYSA-N Synonym: 4-bromo-2-hydroxymethyl-6-methoxyphenol,4-bromo-2-hydroxymethyl-6-methoxybenzenol,acmc-20amo4,benzenemethanol,5-bromo-2-hydroxy-3-methoxy PubChem CID: 894092 IUPAC Name: 4-bromo-2-(hydroxymethyl)-6-methoxyphenol SMILES: COC1=C(O)C(CO)=CC(Br)=C1