accessibility menu, dialog, popup

UserName

CH3H3COHO

CAS RN 3508-94-9

IUPAC Name: 3-methyl-2-phenylbutanoic acid
Synonyms: 3-methyl-2-phenylbutanoic acid
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: HDLQGISFYDYWFJ-UHFFFAOYNA-N
SMILES: CC(C)C(C(O)=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results