accessibility menu, dialog, popup

UserName

CAS RN 35037-73-1

FFFONCO

CAS RN 35037-73-1

IUPAC Name: 1-isocyanato-4-(trifluoromethoxy)benzene
Synonyms: 1-isocyanato-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 203.12
Molecular Formula: C8H4F3NO2
InChi Key: LGPKFIGMLPDYEA-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(C=C1)N=C=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)