CAS RN 35011-47-3
CAS RN 35011-47-3
IUPAC Name:
2,5,6-triamino-1,4-dihydropyrimidin-4-one; sulfuric acid
Synonyms:
2,5,6-triamino-1H-pyrimidin-4-one; sulfuric acid
Molecular Weight (g/mol):
239.21
Molecular Formula:
C4H9N5O5S
InChi Key:
RSKNEEODWFLVFF-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.NC1=NC(=O)C(N)=C(N)N1
1
–
1
of
1
results
1
2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%
CAS: 35011-47-3 Molecular Formula: C4H9N5O5S Molecular Weight (g/mol): 239.206 MDL Number: MFCD00012785 InChI Key: RSKNEEODWFLVFF-UHFFFAOYSA-N Synonym: 2,4,5-triamino-6-hydroxypyrimidine sulfate,6-hydroxy-2,4,5-triaminopyrimidine sulfate,2,5,6-triaminopyrimidin-4 3h-one sulfate,unii-9sz7zx1h0l,sulfuric acid; triaminopyrimidin-4-ol,2,5,6-triamino-4-pyrimidinol sulfate,4 1h-pyrimidinone, 2,5,6-triamino-, sulfate,2,5,6-triaminopyrimidin-4 1h-one sulphate,2,5,6-triaminopyrimidin-4-ol sulfate,9sz7zx1h0l PubChem CID: 296856 IUPAC Name: sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one SMILES: C1(=C(NC(=NC1=O)N)N)N.OS(=O)(=O)O