accessibility menu, dialog, popup

UserName

CAS RN 350-40-3

H3CCH2F

CAS RN 350-40-3

IUPAC Name: 1-fluoro-4-(prop-1-en-2-yl)benzene
Synonyms: 1-fluoro-4-(prop-1-en-2-yl)benzene
Molecular Weight (g/mol): 136.17
Molecular Formula: C9H9F
InChi Key: VIXHMBLBLJSGIB-UHFFFAOYSA-N
SMILES: CC(=C)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results