accessibility menu, dialog, popup

UserName

CAS RN 34966-52-4

H3COOOClCl

CAS RN 34966-52-4

IUPAC Name: ethyl 2-(3,4-dichlorophenyl)-2-oxoacetate
Synonyms: ethyl 2-(3,4-dichlorophenyl)-2-oxoacetate
Molecular Weight (g/mol): 247.07
Molecular Formula: C10H8Cl2O3
InChi Key: NWUGRKGSCYOUFF-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C1=CC(Cl)=C(Cl)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results