accessibility menu, dialog, popup

UserName

CAS RN 349550-81-8

IUPAC Name: 2,3-dihydro-1,4-benzodioxine-5-carboxamide
Synonyms: 2,3-dihydro-1,4-benzodioxine-5-carboxamide
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: OIYLTYTXMKZKCR-UHFFFAOYSA-N
SMILES: NC(=O)C1=C2OCCOC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results