accessibility menu, dialog, popup

UserName

CAS RN 349096-36-2

IUPAC Name: N-(2-ethoxyphenyl)-3-methylbenzamide
Synonyms: N-(2-ethoxyphenyl)-3-methylbenzamide
Molecular Weight (g/mol): 255.32
Molecular Formula: C16H17NO2
InChi Key: WVKNKVSKPRDADI-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1NC(=O)C1=CC(C)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results