accessibility menu, dialog, popup

UserName

FSClOO

CAS RN 349-88-2

IUPAC Name: 4-fluorobenzene-1-sulfonyl chloride
Synonyms: 4-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 194.6
Molecular Formula: C6H4ClFO2S
InChi Key: BFXHJFKKRGVUMU-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)