accessibility menu, dialog, popup

UserName

OOSFFF

CAS RN 349-83-7

IUPAC Name: 2-{[3-(trifluoromethyl)phenyl]sulfanyl}acetate
Synonyms: 2-{[3-(trifluoromethyl)phenyl]sulfanyl}acetate
Molecular Weight (g/mol): 235.2
Molecular Formula: C9H6F3O2S
InChi Key: YBUZSRSMQIEFBR-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC1=CC=CC(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results