accessibility menu, dialog, popup

UserName

CAS RN 348640-06-2

HNNBr

CAS RN 348640-06-2

IUPAC Name: 4-bromo-1H-pyrrolo[2,3-b]pyridine
Synonyms: 4-bromo-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: LEZHTYOQWQEBLH-UHFFFAOYSA-N
SMILES: BrC1=C2C=CNC2=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results