accessibility menu, dialog, popup

UserName

CAS RN 34841-35-5

CH3OCl

CAS RN 34841-35-5

IUPAC Name: 1-(3-chlorophenyl)propan-1-one
Synonyms: 1-(3-chlorophenyl)propan-1-one
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: PQWGFUFROKIJBO-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)