accessibility menu, dialog, popup

UserName

BrSe

CAS RN 34837-55-3

IUPAC Name: (phenylselanyl)bromane
Synonyms: (phenylselanyl)bromane
Molecular Weight (g/mol): 235.98
Molecular Formula: C6H5BrSe
InChi Key: LCEFEIBEOBPPSJ-UHFFFAOYSA-N
SMILES: Br[Se]C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)