accessibility menu, dialog, popup

UserName

CAS RN 34784-05-9

BrN

CAS RN 34784-05-9

IUPAC Name: 6-bromoisoquinoline
Synonyms: 6-bromoisoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: ZTEATMVVGQUULZ-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=NC=CC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results