accessibility menu, dialog, popup

UserName

FOFOFFFOFFFOFFFOFFFFFFFFFFFFFFFFFFF

CAS RN 34761-47-2

IUPAC Name: 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis(trifluoromethyl)-3,6,9,12-tetraoxapentadecanoyl fluoride
Synonyms: 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-heptadecafluoro-2,5,8,11-tetrakis(trifluoromethyl)-3,6,9,12-tetraoxapentadecanoyl fluoride
Molecular Weight (g/mol): 830.11
Molecular Formula: C15F30O5
InChi Key: QDHLRGOEEBWEIB-UHFFFAOYNA-N
SMILES: FC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results