CAS RN 34725-61-6
CAS RN 34725-61-6
IUPAC Name:
sodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Synonyms:
sodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1λ⁶-benzoxathiol-3-yl]benzenolate
Molecular Weight (g/mol):
691.94
Molecular Formula:
C19H9Br4NaO5S
InChi Key:
KGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES:
[Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
1
–
7
of
7
results
1
Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS reagent
CAS: 34725-61-6 Molecular Formula: C19H9Br4NaO5S Molecular Weight (g/mol): 691.94 MDL Number: MFCD00013793 InChI Key: KGANHHDZDYZJEM-UHFFFAOYNA-M Synonym: 3',3'',5',5''-Tetrabromophenolsulfonephthalein PubChem CID: 3791690 IUPAC Name: sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
2
Thermo Scientific Chemicals Bromophenol Blue sodium salt
CAS: 34725-61-6 Molecular Formula: C19H9Br4NaO5S Molecular Weight (g/mol): 691.96 MDL Number: MFCD00013793 Synonym: Bromphenol Blue, water soluble
3
Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS
CAS: 34725-61-6 MDL Number: MFCD00013793
5
Bromophenol Blue Sodium Salt Indicator, Honeywell Fluka™
CAS: 34725-61-6 Molecular Formula: C19H9Br4NaO5S MDL Number: MFCD00013793
6
Bromophenol Blue sodium salt, 0.04% w/v aq. soln.
CAS: 34725-61-6 Molecular Weight (g/mol): 691.96 MDL Number: MFCD00013793