CAS RN 347-84-2
IUPAC Name:
1-(4-fluorophenyl)-2-phenylethan-1-one
Synonyms:
1-(4-fluorophenyl)-2-phenylethanone
Molecular Weight (g/mol):
214.24
Molecular Formula:
C14H11FO
InChi Key:
YFYKGCQUWKAFLW-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1
1
–
3
of
3
results
1
Benzyl 4-Fluorophenyl Ketone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzyl 4-Fluorophenyl Ketone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Benzyl 4-Fluorophenyl Ketone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.