accessibility menu, dialog, popup

UserName

CAS RN 34683-73-3

ClI

CAS RN 34683-73-3

IUPAC Name: 1-chloro-6-iodohexane
Synonyms: 1-chloro-6-iodohexane
Molecular Weight (g/mol): 246.52
Molecular Formula: C6H12ClI
InChi Key: QTJHNJCILMMRIQ-UHFFFAOYSA-N
SMILES: ClCCCCCCI
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)