accessibility menu, dialog, popup

UserName

CAS RN 3468-53-9

OON

CAS RN 3468-53-9

IUPAC Name: phenyl pyridine-3-carboxylate
Synonyms: phenyl pyridine-3-carboxylate
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: QEJPOSAIULNDLU-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=CC=C1)C1=CC=CN=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)