accessibility menu, dialog, popup

UserName

CAS RN 346721-67-3

CH3OHNSOOBrOCH3Cl

CAS RN 346721-67-3

IUPAC Name: 4-bromo-N-(4-chloro-2,5-dimethoxyphenyl)benzene-1-sulfonamide
Synonyms: 4-bromo-N-(4-chloro-2,5-dimethoxyphenyl)benzenesulfonamide
Molecular Weight (g/mol): 406.68
Molecular Formula: C14H13BrClNO4S
InChi Key: WBKSJEMNFRXWFA-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=C(OC)C=C1NS(=O)(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results