accessibility menu, dialog, popup

UserName

H3CNOHNClNCH3O

CAS RN 34654-81-4

IUPAC Name: 6-[(3-chloropropyl)amino]-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 6-[(3-chloropropyl)amino]-1,3-dimethylpyrimidine-2,4-dione
Molecular Weight (g/mol): 231.68
Molecular Formula: C9H14ClN3O2
InChi Key: RPYBDDBZRQGARJ-UHFFFAOYSA-N
SMILES: CN1C(=O)C=C(NCCCCl)N(C)C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results