accessibility menu, dialog, popup

UserName

NNClClHClHCl

CAS RN 346451-15-8

IUPAC Name: 1,4-bis[(4-chlorophenyl)(phenyl)methyl]piperazine dihydrochloride
Synonyms: 1,4-bis[(4-chlorophenyl)(phenyl)methyl]piperazine dihydrochloride
Molecular Weight (g/mol): 560.38
Molecular Formula: C30H30Cl4N2
InChi Key: DLEQNCKNBPYHEY-UHFFFAOYNA-N
SMILES: Cl.Cl.ClC1=CC=C(C=C1)C(N1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results