accessibility menu, dialog, popup

UserName

CAS RN 3458-72-8

NH4NH4NH4OHOOOOOO

CAS RN 3458-72-8

IUPAC Name: triammonium 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: triammonium citrate ammonium citrate tribasic citric acid triammonium salt
Molecular Weight (g/mol): 243.22
Molecular Formula: C6H17N3O7
InChi Key: YWYZEGXAUVWDED-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].[NH4+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (5)
Grade 
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)