CAS RN 34574-69-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxydodecanedioic Acid, TRC
CAS: 34574-69-1 Molecular Formula: C12H22O5 Molecular Weight (g/mol): 246.3 SMILES: O=C(O)CC(O)CCCCCCCCC(O)=O