accessibility menu, dialog, popup

UserName

ONOFN

CAS RN 3456-75-5

IUPAC Name: 2-(5-fluoro-2-nitrophenyl)acetonitrile
Synonyms: 2-(5-fluoro-2-nitrophenyl)acetonitrile
Molecular Weight (g/mol): 180.14
Molecular Formula: C8H5FN2O2
InChi Key: YETOJTGGLXHUCS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C=C1CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results