CAS RN 34523-34-7
IUPAC Name:
4-[1-(hydroxyamino)ethylidene]cyclohexa-2,5-dien-1-one
Synonyms:
4-[1-(hydroxyamino)ethylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol):
151.17
Molecular Formula:
C8H9NO2
InChi Key:
NOGIFJNJJZFUDN-UHFFFAOYSA-N
SMILES:
CC(NO)=C1C=CC(=O)C=C1
1
–
2
of
2
results
1
4’-Hydroxyacetophenone Oxime, TRC
CAS: 34523-34-7 Molecular Formula: C8 H9 N O2 Molecular Weight (g/mol): 151.16 Synonym: 1-(4-Hydroxyphenyl)ethanone oxime,Paracetamol Imp. G (EP),4-Hydroxyacetophenone oxime,Ethanone, 1-(4-hydroxyphenyl)-, oxime,Acetophenone, 4'-hydroxy-, oxime (7CI),1-(4-Hydroxyphenyl)-1-ethanone oxime,4'-Hydroxyacetophenone oxime,NSC 77904,p-Hydroxyacetophenone oxime IUPAC Name: 4-[(Z)-N-hydroxy-C-methylcarbonimidoyl]phenol SMILES: C\C(=N\O)\c1ccc(O)cc1
2
1-(4-Hydroxyphenyl)ethanone Oxime (4-Hydroxyacetophenone Oxime), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.