CAS RN 34422-12-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Elenolic Acid (>90%), TRC
CAS: 34422-12-3 Molecular Formula: C11 H14 O6 Molecular Weight (g/mol): 242.23 Synonym: 2H-Pyran-4-acetic acid, 3-formyl-3,4-dihydro-5-(methoxycarbonyl)-2-methyl-, (2S,3S,4S)-,2H-Pyran-4-acetic acid, 3-formyl-3,4-dihydro-5-(methoxycarbonyl)-2-methyl-, [2S-(2α,3α,4β)]-,Elenolic acid (6CI,7CI),Glutaric acid, 3-(1-formylpropenyl)-2-(hydroxymethylene)-, 1-methyl ester (8CI),(2S,3S,4S)-3-Formyl-3,4-dihydro-5-(methoxycarbonyl)-2-methyl-2H-pyran-4-acetic acid,Elenaic acid IUPAC Name: 2-[(2S,3S,4S)-3-formyl-5-methoxycarbonyl-2-methyl-3,4-dihydro-2H-pyran-4-yl]acetic acid SMILES: COC(=O)C1=CO[C@@H](C)[C@@H](C=O)[C@@H]1CC(=O)O