accessibility menu, dialog, popup

UserName

CH3OHNFFF

CAS RN 344-62-7

IUPAC Name: N-[2-(trifluoromethyl)phenyl]acetamide
Synonyms: O-CF3 acetanilide
Molecular Weight (g/mol): 203.16
Molecular Formula: C9H8F3NO
InChi Key: OXDTZGRSCDEKGO-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results