accessibility menu, dialog, popup

UserName

CAS RN 34369-07-8

IUPAC Name: ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(phosphonatooxy)phosphinate
Synonyms: atp adenosine 5'-triphosphate(4-)
Molecular Weight (g/mol): 503.15
Molecular Formula: C10H12N5O13P3
InChi Key: ZKHQWZAMYRWXGA-KQYNXXCUSA-J
SMILES: NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Molecular Weight (g/mol) 
  • (9)
  • (5)
Quantity 
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (5)
  • (2)
  • (5)
Percent Purity 
  • (7)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (9)
  • (5)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (5)
  • (7)
  • (2)