accessibility menu, dialog, popup

UserName

CH3NNCl

CAS RN 3435-25-4

IUPAC Name: 4-chloro-6-methylpyrimidine
Synonyms: 4-chloro-6-methylpyrimidine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: MVAXKFAQKTWRAH-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=NC=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)