accessibility menu, dialog, popup

UserName

NH3(R)SOO

CAS RN 34317-61-8

IUPAC Name: (2R)-2-azaniumyl-3-(phenylsulfanyl)propanoate
Synonyms: (2R)-2-ammonio-3-(phenylsulfanyl)propanoate
Molecular Weight (g/mol): 197.25
Molecular Formula: C9H11NO2S
InChi Key: XYUBQWNJDIAEES-QMMMGPOBSA-N
SMILES: [NH3+][C@@H](CSC1=CC=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)