CAS RN 3430-10-2
IUPAC Name:
2-methylpyridin-3-amine
Synonyms:
2-methylpyridin-3-amine
Molecular Weight (g/mol):
108.14
Molecular Formula:
C6H8N2
InChi Key:
ZSFPJJJRNUZCEV-UHFFFAOYSA-N
SMILES:
CC1=C(N)C=CC=N1
1
–
2
of
2
results
2
3-Amino-2-methylpyridine, 97%, Thermo Scientific Chemicals
CAS: 3430-10-2 MDL Number: MFCD03788195