CAS RN 34271-54-0
CAS RN 34271-54-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DL-Theanine, TRC
CAS: 34271-54-0 Molecular Formula: C7H14N2O3 Molecular Weight (g/mol): 174.2 Synonym: Glutamine, N-ethyl- (9CI, ACI),DL-Glutamine, N-ethyl- (ZCI),Glutamine, N-ethyl-, DL- (8CI),N-Ethylglutamine (ACI),2-Amino-5-(ethylamino)-5-oxopentanoic acid,DL-Theanine,N-Ethyl-DL-glutamine IUPAC Name: 2-amino-5-(ethylamino)-5-oxopentanoic acid SMILES: CCNC(=O)CCC(N)C(=O)O