accessibility menu, dialog, popup

UserName

CAS RN 342405-27-0

CH3NNOH

CAS RN 342405-27-0

IUPAC Name: (4-methyl-2-phenylpyrimidin-5-yl)methanol
Synonyms: (4-methyl-2-phenylpyrimidin-5-yl)methanol
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: LXJHDOHHZSZIMJ-UHFFFAOYSA-N
SMILES: CC1=NC(=NC=C1CO)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results