accessibility menu, dialog, popup

UserName

CAS RN 34014-18-1

H3CNHONCH3SNNCH3H3CCH3

CAS RN 34014-18-1

IUPAC Name: 1-(5-tert-butyl-1,3,4-thiadiazol-2-yl)-1,3-dimethylurea
Synonyms: tebuthiuron
Molecular Weight (g/mol): 228.31
Molecular Formula: C9H16N4OS
InChi Key: HBPDKDSFLXWOAE-UHFFFAOYSA-N
SMILES: CNC(=O)N(C)C1=NN=C(S1)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results