accessibility menu, dialog, popup

UserName

CAS RN 34006-60-5

H3COOOCH3Cl

CAS RN 34006-60-5

IUPAC Name: ethyl 2-(1-chloroethoxy)acetate
Synonyms: ethyl 2-(1-chloroethoxy)acetate
Molecular Weight (g/mol): 166.6
Molecular Formula: C6H11ClO3
InChi Key: WDCHMODANTVRTF-UHFFFAOYNA-N
SMILES: CCOC(=O)COC(C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results