accessibility menu, dialog, popup

UserName

H3COOHNOHHCl

CAS RN 33996-30-4

IUPAC Name: ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride
Synonyms: ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride
Molecular Weight (g/mol): 195.64
Molecular Formula: C7H14ClNO3
InChi Key: HHXSZDXMSRXWJV-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C1CC(O)CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)