CAS RN 33939-97-8
CAS RN 33939-97-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Aluminum lanthanum isopropoxide, 99% (metals basis), 7% w/v in isopropanol, Thermo Scientific™
CAS: 33939-97-8 MDL Number: MFCD00143591 Synonym: Lanthanum aluminum isopropoxide