CAS RN 339071-18-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(3-Chloro-4-morpholinophenyl)-N'-hydroxyformimidamide, MedChemExpress
MedChemExpress TS-011 is a selective inhibitor of 20-Hydroxyeicosatetraenoic acid synthesis.