accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3COCH3

CAS RN 338998-93-9

IUPAC Name: 4,4,5,5-tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 208.06
Molecular Formula: C11H17BO3
InChi Key: FNPZFZKLYGWKLH-UHFFFAOYSA-N
SMILES: CC1=CC=C(O1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results