accessibility menu, dialog, popup

UserName

ONS

CAS RN 33898-90-7

IUPAC Name: 3-oxo-3-(thiophen-2-yl)propanenitrile
Synonyms: 3-oxo-3-(thiophen-2-yl)propanenitrile
Molecular Weight (g/mol): 151.18
Molecular Formula: C7H5NOS
InChi Key: XWWUQBHVRILEPB-UHFFFAOYSA-N
SMILES: O=C(CC#N)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results