accessibility menu, dialog, popup

UserName

CAS RN 3385-34-0

IUPAC Name: ethyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate
Synonyms: ethyl 2,4-dimethyl-6-oxopyran-3-carboxylate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: FBPWNVQUVXSXKS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)OC(=O)C=C1C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)