accessibility menu, dialog, popup

UserName

ONFFFClSOOCl

CAS RN 338422-71-2

IUPAC Name: 4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzene-1-sulfonyl chloride
Synonyms: 4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulfonyl chloride
Molecular Weight (g/mol): 372.14
Molecular Formula: C12H6Cl2F3NO3S
InChi Key: NEQIUPPXYAOTSF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results